Tisk…
Absorbent mat pads for viscous liquids, PIG®
Dodavatel: NEW PIG
MAT209
NEWPMAT209EA 8480 CZK
NEWPMAT209
Absorbent mat pads for viscous liquids, PIG®
Absorpční materiály
White polyester fibre absorbent mat pads for viscous liquids for picking up paint, glue, resins, shampoo, liquid chocolate, syrup, varnish.
Up to 3,63 litres of viscous fluids per pad.