Tisk…
High-visibility safety floor runners, PIG®
Dodavatel: NEW PIG
GRPSB36200-BK GRPSB36201-BK
NEWPGRPSB36200-BKEA 13740 CZK
NEWPGRPSB36200-BK NEWPGRPSB36201-BK
High-visibility safety floor runners, PIG®
Rohože Rohože lepící
Provide a no-slip path in hazardous walking areas with our super-safe, adhesive-backed absorbent runner - its hi-viz yellow safety edges can't be missed.
  • Tapes and paints are easy to see but do nothing to create a safer floor surface
  • The PIG® high-visibility safety floor runner is the only floor safety solution that combines a high-traction walking surface with a hi-viz safety edge pattern
  • Adhesive backing keeps mat exactly where you put it with no shifting, bunching up or flipping over; traps dirt, salt and moisture to keep it from being tracked around
More Product Information