Tisk…
Nalgene® Biotainer® Closures for Bottles, InVitro™
Dodavatel: Thermo Fisher Scientific
2560-0489 2560-0490
NALG2560-0489EA 7040 CZK
NALG2560-0489 NALG2560-0490
Nalgene® Biotainer® Closures for Bottles, InVitro™
Uzávěry Uzávěry k láhvím
Designed to aseptically handle solutions in biopharmaceutical, biotechnology and laboratory liquid transfer applications, these two- and three-ported closures are designed for Thermo Scientific™ Nalgene™ Biotainer™ bottles and carboys. With Animal Derived Component-Free (ADCF) components, including seals that are sonically-welded to the cap, and leak-proof closures, security of high value contents are guaranteed during fluid transfer.
  • With ADCF components
  • Including seals that are sonically welded to the cap
  • Leak-proof closures
  • Security of high value
More Product Information