Tisk…
Muffle furnaces with lift up door, Tmax 1300 °C and standard controller B510, LT 5/13 - LT 15/13
Dodavatel: NABERTHERM
L-055H3ANL L-055H3ANLG L-055H3ANLGT L-055H3ANLGTZ L-055H3ANLGZ L-055H3ANLT L-055H3ANLTZ L-055H3ANLZ
NABEL-055H3ANLEA 155950 CZK
NABEL-055H3ANL NABEL-055H3ANLG NABEL-055H3ANLGT NABEL-055H3ANLGTZ NABEL-055H3ANLGZ NABEL-055H3ANLT NABEL-055H3ANLTZ NABEL-055H3ANLZ
Muffle furnaces with lift up door, Tmax 1300 °C and standard controller B510, LT 5/13 - LT 15/13
Sušárny Pece vysokoteplotní
These furnaces feature heating elements on supporting tubes that radiate freely into the chamber, ensuring particularly short heating times. With the robust lightweight refractory brick insulation, they can reach a max. working temperature of 1300 °C.
  • Heating from two sides
  • Multi-layer insulation with robust lightweight refractory bricks in the furnace chamber
  • Textured stainless steel housing
  • Dual shell housing for low external temperatures and high stability
  • Lift up door (LT) with hot surface facing away from the operator
  • Exhaust air outlet on rear wall
More Product Information