Tisk…
Safety Storage Cabinet, Type 90, ION-PRO-90 STORE model IO90.195.120.PS.WDC
Dodavatel: asecos
41429047 41386 41387 41388 41389
135-2881EA 178840 CZK
135-2881 MSGA41386 MSGA41387 MSGA41388 MSGA41389
Safety Storage Cabinet, Type 90, ION-PRO-90 STORE model IO90.195.120.PS.WDC
Nábytek kancelářský Skříně pro zásoby
Safe passive storage of lithium-ion batteries.
  • Robust construction and longevity; Convenient cabinet entry
  • Integrated potential-free alarm contact; No unauthorised use
  • Integrated transport base; Easy alignment
  • Smoke detector and temperature sensor; Pressure relief
  • All-around protection
  • LED display
  • Fire suppression system
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface.
Convenient cabinet entry: open both doors at the same time with just one hand. Integrated potential-freeMore Product Information