Tisk…
Cabinets for chemicals, CF-CLASSIC
Dodavatel: asecos
30802 30801 30800 30799 30798 30797 30796
MSGA30802EA 24550 CZK
MSGA30802 MSGA30801 MSGA30800 MSGA30799 MSGA30798 MSGA30797 MSGA30796
Cabinets for chemicals, CF-CLASSIC
Nábytek kancelářský Skříně pro zásoby
Safe and approved indoor storage of different hazardous substances in one safety storage cabinet.
  • Robust construction and longevity – sturdy sheet-steel construction
  • No unauthorised use: door lockable with cylinder locking (integrated in the turning handle)
  • Ventilation – ventilation openings in the door, ready for connection (DN 75) on the top to a technical exhaust system
  • Height adjustable, galvanised tray shelves, load capacity 50 kg
Made in Germany.
Informace pro objednávání: All cabinets are supplied with tray shelves (galvanized sheet steel)More Product Information