Tisk…
Disposable earplugs, Pura-Fit®
Dodavatel: Moldex
7750 6900 7700
MOLD7750EA 3650 CZK
MOLD7750 MOLD6900 MOLD7700
Disposable earplugs, Pura-Fit®
Chrániče zátkové Chrániče zátkové jednorázové
These plugs made of soft PU foam provide protection against higher noise levels.
  • Tapered form makes it easy to insert the plugs in the ear
  • Outer surface is smooth, so it doesn't irritate the canal
  • Bright colour for improved visibility
SNR: 36 dB
Certifikace: EN 352-2:2002
Balení: As pairs or in the hygienic dispenser system MoldexStation.
Objednat
Produktová specifikace
Environmentally Preferable Safer Human and Environmental Health