Tisk…
Heating mantle, KM-ME series
Dodavatel: MOHR
82001000 92010000 92020000 92004000 92006000
MOHR82001000EA 14710 CZK
MOHR82001000 MOHR92010000 MOHR92020000 MOHR92004000 MOHR92006000
Heating mantle, KM-ME series
Ohřívače Hnízda topná
Metal-cased heating mantle with built-in controller for operating temperatures up to 450 °C. Suitable for round bottomed flasks with volume up to 20 L.
  • Flexible glass yarn heating element provides even heat transfer and gentle treatment of the glassware
  • Chemical resistant plastic-coated metal housing
  • Thermally insulated and earthed
  • Predrilled threaded hole for clamp KM-SK
  • Built-in heating zone switch
  • With residual current detection (RCD)
System of protection: Casing IP 43, heating surface IP 00.