Tisk…
Octaheel Baby Blood Lancets
Dodavatel: Vitrex Medical A/S
335313 334313 333313 332313 331313
MODU335313EA 2970 CZK
MODU335313 MODU334313 MODU333313 MODU332313 MODU331313
Octaheel Baby Blood Lancets
Lancety
Vitrex Octaheel Baby are sterile cutting safety lancets intended for high volume blood flow with minimum depth of penetration.
  • Activation button is secured by safety tab
  • Non-detachable safety tab
  • Blade is fully shielded before and after use which provides user safety
  • Anti-slip texture and one hand operation available for easier handling
  • Pre-loaded, button activated design
  • Single-use only, sterilised by gamma irradiation
  • Ultra sharp blade with high speed cutting securing minimal trauma and fast healing
  • Narrowed lancing slot helps for easier positioning