Tisk…
Oil sorbents
Dodavatel: 3M
HP156 HP100 T150 TF2001
MMMAHP156EA 9170 CZK
MMMAHP156 MMMAHP100 MMMAT150 MMMATF2001
Oil sorbents
Absorpční materiály
Oil adsorbent sheets or pads on a roll for spill control.
  • Hydrophobic
  • Oil (not water) is recovered
  • Can be used outside as well as in-plant