Tisk…
Maintenance sorbents
Dodavatel: 3M
MB2001 MA2002
MMMAMB2001EA 8710 CZK
MMMAMB2001 MMMAMA2002
Maintenance sorbents
Absorpční materiály
For use in production facilities and maintenance shops to handle routine leaks, drips, spray, and fluid splashes from nonaggressive liquids. Nonaggressive liquids are those used in daily operations which do not require the use of personal protective equipment. These include oils, coolants, cutting fluids, aqueous-based liquids, and petroleum-based products.
Unlike clay granules, these sorbents are not messy and can be used on equipment. This allows placement at the source of the leak, preventing spread to floor. The various sizes and styles available also permit use under, around, or forMore Product Information