Tisk…
Powered and supplied air replacement visors and lenses, Versaflo™
Dodavatel: 3M
M927
MMMAM927EA 1950 CZK
MMMAM927
Powered and supplied air replacement visors and lenses, Versaflo™
Polomasky a filtrační respirátory Systémy polomasek
The Versaflo™ coated visor M-927 is a clear, polycarbonate replacement visor designed for use with 3M™ Versaflo™ M-Series Headtops.
  • Coated to provide chemical and scratch resistance
  • Clear polycarbonate
  • CE approved to EN 166 1:BT:3
Replace scratched and worn visors to ensure continued good vision. This replacement polycarbonate visor provides chemical and scratch resistance for 3M™ Versaflo™ M-Series Headtops.