Tisk…
Powered air respirator visor frame assemblies, Versaflo™
Dodavatel: 3M
M-961
MMMAM-961EA 5340 CZK
MMMAM-961
Powered air respirator visor frame assemblies, Versaflo™
Polomasky a filtrační respirátory Systémy polomasek
The Versaflo™ Over-Visor Frame M-961 is a carrier frame designed to fit Versaflo™ Over-Visor M-967 and Versaflo™ radiant heat covers for Versaflo™ M-Series Helmets.
  • Carrier frame to fit over-visor and radiant heat covers to M-Series helmets
  • Designed specifically for use with M-Series helmets
This frame is made from aluminium.