Tisk…
PELTOR™ HY80 Camelback Gel Sealing Rings
Dodavatel: 3M
HY80
MMMAHY80EA 3680 CZK
MMMAHY80
PELTOR™ HY80 Camelback Gel Sealing Rings
Těsnění Těsnící kroužky
Gel ear cushions for use on a wide variety of 3M™ PELTOR™ Headsets. These gel sealing rings features a super-thin polyurethane skin with a double-hump top face design and a silicon gel-filled bladder with a Confor™ foam backing.
  • Super-soft foam for enhanced comfort
  • Sealing rings feature a super-thin polyurethane skin with a double-hump top face design and a silicon gel-filled bladder with a Confor™ foam backing
Gel filled replacement cushions maintain a comfortable, noise-blocking seal around ears. Sealing rings maintain original service performance of Peltor products.