Tisk…
Adflo™ Powered Air Respirator Filter and Battery Kit
Dodavatel: 3M
H837301
MMMAH837301EA 17010 CZK
MMMAH837301
Adflo™ Powered Air Respirator Filter and Battery Kit
Polomasky a filtrační respirátory Systémy polomasek
AdFlo™ Powered air respirator filter and battery kit contains the Adflo™ filter and Adflo™ heavy-duty Li-ion battery. It’s designed for use with 3M™ Adflo™ powered air respirator.
  • Replacement powered air respirator and battery
  • Upgrade kit for Adflo™ Powered air respirator
  • Captures organic, inorganic, and acid gases and vapours
  • Always to be used together with particle filter
  • Heavy-duty battery fully charges in 5,5 hours and has an operating time of 10 to 12 hours
Get reliable protection from gases and vapours by upgrading the 3M™ Adflo™ Powered AirMore Product Information