Tisk…
Multi-purpose microfibre wipes, 2030 series, Scotch-Brite™
Dodavatel: 3M
2030GN 2030RT 2030GB
MMMA2030GNEA 0 CZK
MMMA2030GN MMMA2030RT MMMA2030GB
Multi-purpose microfibre wipes, 2030 series, Scotch-Brite™
Ubousky/Utěrky Utěrky čistící pro opakovatelné použití Utěrky pro běžné aplikace
The 2030 series Scotch-Brite™ multi purpose microfibre wipes can be used dry or damp to effectively remove oil, grease, dirt, water, and dust from all common cleaning surfaces. They're soft and flexible, and designed to resist autoclave.
  • Easy to rinse clean
  • Eliminates the need for cleaning agents in most cases
  • Machine washable up to 200 times at 95 °C
Efficiently absorb dust, dirt, fingerprints, grease, oil, and moisture from surfaces with Scotch-Brite™ Multi Purpose Microfibre Wipes, 2030 series. These microfibre wipes are soft and flexible, with aMore Product Information