Tisk…
Microtiter tubes system
Dodavatel: Thermo Fisher Scientific
3487 3492 3496 3425-11 3426-11
MLBP3487EA 0 CZK
MLBP3487 MLBP3492 MLBP3496 MLBP3425-11 MLBP3426-11
Microtiter tubes system
Zkumavky Mikrozkumavky
Tubes made of PP, natual, caps made of PE, natural.
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight plug strips can be easily cut apart for individual samples
Microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage. Frosted writing areas on each tube and an alphanumeric rack simplify organization andMore Product Information