Tisk…
Centrifugal filter units, Amicon® Ultra-15
Dodavatel: MILLIPORE
UFC901008 UFC901024 UFC901096 UFC903008 UFC903024 UFC903096 UFC910008 UFC910024 UFC910096 UFC905008 UFC905024 UFC900308 UFC900324 UFC905096 UFC900396
MILYUFC901008EA 3590 CZK
MILYUFC901008 MILYUFC901024 MILYUFC901096 MILYUFC903008 MILYUFC903024 MILYUFC903096 MILYUFC910008 MILYUFC910024 MILYUFC910096 MILYUFC905008 MILYUFC905024 MILYUFC900308 MILYUFC900324 MILYUFC905096 MILYUFC900396
Centrifugal filter units, Amicon® Ultra-15
Filtry Filtry centrifugační
These centrifugal filter units can be used for a number of applications: concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; purification of macromolecular components found in tissue culture extracts or cell lysates and protein removal prior to HPLS; concentration of dilute or pre-purified proteins from column eluents; desalting, buffer exchange and protein dialysis; size exclusion and protein fractionation.
  • High retentate recovery of >90%; 100% integrity tested
  • Vertical membrane reduces
More Product Information