Tisk…
Millex® Syringe Filter Units, Hydrophilic PTFE Membrane
Dodavatel: MILLIPORE
SLLGC13NL SLCRT13NL SLLGC25NS SLLHC25NS SLLGH25NS SLLGH13NK SLLGH13NL SLLHC13NL SLLGH25NB SLCR025NS SLLHR04NK
MILYSLLGC13NLEA 9180 CZK
MILYSLLGC13NL MILYSLCRT13NL MILYSLLGC25NS MILYSLLHC25NS MILYSLLGH25NS MILYSLLGH13NK MILYSLLGH13NL MILYSLLHC13NL MILYSLLGH25NB MILYSLCR025NS MILYSLLHR04NK
Millex® Syringe Filter Units, Hydrophilic PTFE Membrane
Filtry Filtry stříkačkové
HPLC certified filters for the filtration of organic and aqueous solutions in HPLC.
  • Low protein binding and high chemical resistance
Tested and certified for UV-absorbing substances at wavelengths of 210 and 254 nm with water and acetonitrile; ensuring consistent quality between batches.
Inlet/outlet: Luer lock female/Luer male.