Tisk…
Millex® Syringe Filters, HPF, Nylon Membrane with Glass Fibre Pre-filter
Dodavatel: MILLIPORE
SLHNMZ5NZ SLHNM25NS SLGNM25NS
MILYSLHNMZ5NZEA 31810 CZK
MILYSLHNMZ5NZ MILYSLHNM25NS MILYSLGNM25NS
Millex® Syringe Filters, HPF, Nylon Membrane with Glass Fibre Pre-filter
Filtry Filtry stříkačkové
These filters include multiple media: a graduated prefilter and a membrane filter for high throughput filtration of viscous or particle-laden solutions.
  • High-density polyethylene housing compatible with the most commonly used solvents
  • Multi-layer filter can improve throughput of high particulate/viscous samples
Applications include analytical sample preparation, wine analysis and general particulate removal.