Tisk…
Ultracel PL Ultrafiltration Membrane Disc Filters
Dodavatel: MILLIPORE
PLAC02510 PLAC04310 PLAC04710 PLAC06210 PLAC07610 PLAC09005 PLAC15005 PLBC02510 PLBC04310 PLBC04710 PLBC06210 PLBC07610 PLBC09005 PLBC15005 PLCC02510 PLCC04310 PLCC04710 PLCC06210 PLCC07610 PLCC09005 PLCC15005 PLGC02510 PLGC04310 PLGC04710 PLGC06210 PLGC07610 PLGC09005 PLHK04310 PLHK04710 PLHK06210 PLHK07610 PLHK09005 PLTK02510 PLTK04310 PLTK04710 PLTK06210 PLTK09005 PLTK15005 PLTK07610 PLGC15005 PLHK02510 PLHK15005
MILYPLAC02510EA 4600 CZK
MILYPLAC02510 MILYPLAC04310 MILYPLAC04710 MILYPLAC06210 MILYPLAC07610 MILYPLAC09005 MILYPLAC15005 MILYPLBC02510 MILYPLBC04310 MILYPLBC04710 MILYPLBC06210 MILYPLBC07610 MILYPLBC09005 MILYPLBC15005 MILYPLCC02510 MILYPLCC04310 MILYPLCC04710 MILYPLCC06210 MILYPLCC07610 MILYPLCC09005 MILYPLCC15005 MILYPLGC02510 MILYPLGC04310 MILYPLGC04710 MILYPLGC06210 MILYPLGC07610 MILYPLGC09005 MILYPLHK04310 MILYPLHK04710 MILYPLHK06210 MILYPLHK07610 MILYPLHK09005 MILYPLTK02510 MILYPLTK04310 MILYPLTK04710 MILYPLTK06210 MILYPLTK09005 MILYPLTK15005 MILYPLTK07610 MILYPLGC15005 MILYPLHK02510 MILYPLHK15005
Ultracel PL Ultrafiltration Membrane Disc Filters
Filtry Filtry membránové
Ultrafiltration cut disc membranes are available in Ultracel® regenerated cellulose and Biomax® polyethersulvone (PES).
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic and tight microstructure assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.