Tisk…
Aerosol analysis monitors
Dodavatel: MILLIPORE
AP1002500 MAWP025A0 MAWP025AC
MILYAP1002500EA 1680 CZK
MILYAP1002500 MILYMAWP025A0 MILYMAWP025AC
Aerosol analysis monitors
Filtry Kazety filtrační
The PS monitor is used for sample collection and particle analysis. The PP monitor is used for microscopic particle analysis and sample collection for HPLC and UV detection.
Membrane: mixed cellulose esters with cellulose support pad; housing: PS or carbon filled PP.
Objednat
Produktová specifikace
Environmentally Preferable
Velikost póru
Materiál
Filtrační plocha
Ø
Přípojka pro příkon/výkon spojení