Tisk…
Membrane filters, Isopore™
Dodavatel: Merck
TETP14250 GTTP04700 TSTP02500 VCTP04700 ATTP01300 ATTP03700 ATTP14250 DTTP01300 DTTP04700 GTBP01300 GTTP01300 GTTP03700 GTTP09030 GTTP14250 HTBP01300 HTTP03700 HTTP09030 HTTP14250 RTTP01300 RTTP14250 TCTP01300 TCTP02500 TCTP14250 TETP01300 TETP02500 TETP04700 TMTP01300 TMTP09030 TMTP14250 TSTP01300 TSTP14250 TTTP02500 VCTP01300 VCTP02500 VCTP14250 GTTP02500 GTBP02500 GTBP04700 HTTP02500 HTTP04700 HTBP02500 HTBP04700 ATTP02500 ATTP04700 RTTP02500 RTTP04700 TSTP04700 TMTP02500 TMTP04700 HTTP01300 TTTP04700 VCTP07625 TMTP0190R DTTP02500 TCTP04700
MILXTETP14250EA 0 CZK
MILXTETP14250 MILYGTTP04700 MILYTSTP02500 MILYVCTP04700 MILYATTP01300 MILYATTP03700 MILYATTP14250 MILYDTTP01300 MILYDTTP04700 MILYGTBP01300 MILYGTTP01300 MILYGTTP03700 MILYGTTP09030 MILYGTTP14250 MILYHTBP01300 MILYHTTP03700 MILYHTTP09030 MILYHTTP14250 MILYRTTP01300 MILYRTTP14250 MILYTCTP01300 MILYTCTP02500 MILYTCTP14250 MILYTETP01300 MILYTETP02500 MILYTETP04700 MILYTMTP01300 MILYTMTP09030 MILYTMTP14250 MILYTSTP01300 MILYTSTP14250 MILYTTTP02500 MILYVCTP01300 MILYVCTP02500 MILYVCTP14250 MILYGTTP02500 MILYGTBP02500 MILYGTBP04700 MILYHTTP02500 MILYHTTP04700 MILYHTBP02500 MILYHTBP04700 MILYATTP02500 MILYATTP04700 MILYRTTP02500 MILYRTTP04700 MILYTSTP04700 MILYTMTP02500 MILYTMTP04700 MILYHTTP01300 MILYTTTP04700 MILYVCTP07625 MILYTMTP0190R MILYDTTP02500 MILYTCTP04700
Membrane filters, Isopore™
Filtry Filtry membránové
These PC hydrophilic membrane are track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. They are composed of polycarbonate film, which has a smooth, glass like surface for clearer sample observation.
  • Membrane for microscopy and visual analysis
  • Membrane structure retains particles on the surface, simplifying counting and analysis
  • Do not stain, resulting in low
More Product Information