Tisk…
Membrane filters, Durapore®
Dodavatel: MILLIPORE
GVWP04700 GVWP09050 HVLP04700 SVLP04700 VVHP04700 GVWP01300 DVPP01300 DVPP02500 DVPP04700 DVPP09050 DVPP14250 GVHP01300 GVHP02500 GVHP09050 GVHP14250 GVWP10050 HVHP02500 HVHP04700 HVHP09050 HVHP14250 HVLP01300 HVLP14250 VVLP04700 HVLP09050 SVLP02500 SVLP09050 SVWG04700 VVLP01300 VVLP02500 VVLP09050 VVLP14250 GVWP14250 GVHP04700 GVWP02500 HVLP02500 GVHP00010 HVHP01300 VVLP00010 DVPP00010 GVWP00010 DVPP08250 HVLP00010 HVWG04700 SLVP07550 DVPP29325 HVLP08050 SVLP01300
MILYGVWP04700EA 5790 CZK
MILYGVWP04700 MILYGVWP09050 MILYHVLP04700 MILYSVLP04700 MILYVVHP04700 MILYGVWP01300 MILYDVPP01300 MILYDVPP02500 MILYDVPP04700 MILYDVPP09050 MILYDVPP14250 MILYGVHP01300 MILYGVHP02500 MILYGVHP09050 MILYGVHP14250 MILYGVWP10050 MILYHVHP02500 MILYHVHP04700 MILYHVHP09050 MILYHVHP14250 MILYHVLP01300 MILYHVLP14250 MILYVVLP04700 MILYHVLP09050 MILYSVLP02500 MILYSVLP09050 MILYSVWG04700 MILYVVLP01300 MILYVVLP02500 MILYVVLP09050 MILYVVLP14250 MILYGVWP14250 MILYGVHP04700 MILYGVWP02500 MILYHVLP02500 MILYGVHP00010 MILYHVHP01300 MILYVVLP00010 MILYDVPP00010 MILYGVWP00010 MILYDVPP08250 MILYHVLP00010 MILYHVWG04700 MILYSLVP07550 MILYDVPP29325 MILYHVLP08050 MILYSVLP01300
Membrane filters, Durapore®
Filtry Filtry membránové
Durapore® PVDF white membranes provide high flow rates and throughput, low extractables and broad chemical compatibility. Hydrophlic membrane binds far less protein than nylon, nitrocellulose, or PTFE membranes.
  • Can be sterilised by ethylene oxide and gamma irradiation
  • Available in several pore sizes (both hydrophilic and hydrophobic)
  • Have very low protein binding to minimize interaction with your sample and maximise recovery
Filter surface: plain
Bacterial endotoxins: 0,5 EU/ml
Gravimetric extractables: <0,5%