Tisk…
Glass fiber filters without binder resin
Dodavatel: MILLIPORE
AP4001000 AP4002405 AP4003705 AP4004705 AP4007000 AP4009000 AP4014250 APFB02500 APFA04700 APFB04700 APFC04700 APFC09050 APFD04700 APFD09050 APFF04700 APFF09050 APFF02500 AP4002500 AP4004700 APFB03700 APFB15050 APFC03700 APFF12450 APFF14250 AP408X105 APFC02500 APFD02500
MILYAP4001000EA 1680 CZK
MILYAP4001000 MILYAP4002405 MILYAP4003705 MILYAP4004705 MILYAP4007000 MILYAP4009000 MILYAP4014250 MILYAPFB02500 MILYAPFA04700 MILYAPFB04700 MILYAPFC04700 MILYAPFC09050 MILYAPFD04700 MILYAPFD09050 MILYAPFF04700 MILYAPFF09050 MILYAPFF02500 MILYAP4002500 MILYAP4004700 MILYAPFB03700 MILYAPFB15050 MILYAPFC03700 MILYAPFF12450 MILYAPFF14250 MILYAP408X105 MILYAPFC02500 MILYAPFD02500
Glass fiber filters without binder resin
Filtry Filtry ze skleněných mikrovláken
The filters without binder resin retain their structural integrity without weight loss when heated up to 500 °C and can therefore be used in gravimetric analysis as well as for the filtration of hot gases.
  • Can be sterilised by EO, gamma, or autoclave.
Type APFA: Retains fine particles with good efficiency, even at high flow rates, used for monitoring wastewater and collecting suspended particles in gases, also have biochemical applications like collection of cells and filtration of protein or nucleic acid precipitates.
Type APFB: Higher mechanicalMore Product Information
Objednat
Produktová specifikace
Environmentally Preferable
Hmotnost
Tloušťka
Záchyt částic
Velikost póru
Objemový průtok vody
Průtok vzduchu
Maximální teplota