Tisk…

Visualizer and Visualizer EC Western Blot Detection Kits

Dodavatel: MILLIPORE

64-201 64-202BP 64-202 64-201BP 64-202SP 64-301 64-301BP 64-301SP
MILY64-201EA 13660 CZK
MILY64-201 MILY64-202BP MILY64-202 MILY64-201BP MILY64-202SP MILY64-301 MILY64-301BP MILY64-301SP
Visualizer and Visualizer EC Western Blot Detection Kits
Činidla pro elektroforézu Western Blotting Reagents

The Visualizer Western Blot Detection kit is designed for the sensitive detection of proteins in chemiluminescent Western blots.


  • Storage: Stable for 18 months at 4 °C

  • Application: For use on immunoblots


The reagents in this kit have been matched to optimise the range and sensitivity of detection using various antibodies in Western blot assays.

Components: Goat anti-mouse IgG, goat anti-rabbit IgG, HRP conjugate Visualizer detection reagent A Visualizer detection reagent B.



Visualizer EC western blot detection kits are designed with a high glow reaction chemiluminescent substrate with outstanding sensitivity, long-lasting signal and low cost. Visualizer EC offers the highest signal-to-noise ratios of any existing luminol peroxidase formulations, resulting in high sensitivity and low backgrounds.


Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR