Tisk…
Centriprep® Centrifugal Filters
Dodavatel: MILLIPORE
4303 4305 4306 4307 4310 4304
MILY4303EA 0 CZK
MILY4303 MILY4305 MILY4306 MILY4307 MILY4310 MILY4304
Centriprep® Centrifugal Filters
Filtry Filtry centrifugační
These centrifugal filters are disposable ultrafiltration non sterile devices used for purifying, concentrating, and desalting biological samples in the 2 to 15 ml volume range.
  • For use with samples containing high levels of solutes
  • Unique inverse flow mode of operation with large dead stop
  • Fast sample processing
  • Fits standard swinging-bucket rotor for 50 ml tubes
They may also be used for filtration applications. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 ml centrifugeMore Product Information