Tisk…
Ultracel YM Ultrafiltration Membrane Disc Filters
Dodavatel: MILLIPORE
14432 14442AM 14451 14461 14412 14422AM 13651
MILY14432EA 13600 CZK
MILY14432 MILY14442AM MILY14451 MILY14461 MILY14412 MILY14422AM MILY13651
Ultracel YM Ultrafiltration Membrane Disc Filters
Filtry Filtry membránové
Ultrafiltration cut disc membranes are available in Ultracel® regenerated cellulose and Biomax® polyethersulvone (PES).
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic and tight microstructure assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.