Tisk…
Mobile Phase Filters
Dodavatel: MICROSOLV
58846-05
MICS58846-05EA 1390 CZK
MICS58846-05
Mobile Phase Filters
Filtry Filtry pro práci s kapalinami
Biocompatible, last drop, PEEK and polypropylene mobile phase filters. PTFE filter element and will provide very high filter capacity and throughput while using a maximum amount of solvent. The unique design allows for this flat filter to vacuum up to the last drop of liquid in the reservoir bottle.
  • Tubing connector uses the fitting and an ETFE ferrule to hold non-flanged low pressure tubing in the filter without leaking
  • The 5 µm pore size element will filter undissolved particles from your solvents that are 5 µm or larger preventing them from damaging
More Product Information