Tisk…
Masterflex® NSF-Approved Turbine Flowmeters, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX98516-90EA 8150 CZK
MFLX98516-90 MFLX98516-92
Masterflex® NSF-Approved Turbine Flowmeters, Avantor®
Průtokoměry
Designed for low-viscosity flow measurement in water and beverage applications.
  • NSF Standard 61 listed—ideal for use with water sampling and processing applications
  • Designed with 3/8” NPT(M) process connections
  • Suitable for use with 1.8 to 16 cSt liquids
  • Features an accuracy of ±2% of reading
Designed for low-flow OEM and pilot applications, these turbine flowmeters are highly accurate and repeatable and feature a Hall effect sensor for superior accuracy. The sensor’s standard power and output specifications make it easy to retrofit existingMore Product Information