Tisk…
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX96410-13EA 2200 CZK
MFLX96410-13 MFLX96410-14 MFLX96410-16 MFLX96410-17 MFLX96410-18 MFLX96410-25
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Hadičky Hadičky peristaltické pumpy
Excellent biocompatibility that meets multiple classifications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and
More Product Information