Tisk…
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Solva™, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX95606-12EA 1550 CZK
MFLX95606-12 MFLX95606-14 MFLX95606-22 MFLX95606-24 MFLX95606-26 MFLX95606-28 MFLX95606-30 MFLX95606-32 MFLX95606-36 MFLX95606-38 MFLX95606-40 MFLX95606-42 MFLX95606-44 MFLX95606-48 MFLX95606-10 MFLX95606-18 MFLX95606-34 MFLX95606-46
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Solva™, Avantor®
Hadičky Hadičky peristaltické pumpy
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Ideal for chromatography applications and for the transfer of fuels, distillates, lubricants, and glycol-based coolants
  • Resistant to ozone and UV light; resistant to cracking, swelling, and
More Product Information