Tisk…
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Solva™, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX95605-12EA 1510 CZK
MFLX95605-12 MFLX95605-18 MFLX95605-22 MFLX95605-24 MFLX95605-26 MFLX95605-28 MFLX95605-30 MFLX95605-32 MFLX95605-36 MFLX95605-40 MFLX95605-42 MFLX95605-44 MFLX95605-48 MFLX95605-38 MFLX95605-46 MFLX95604-36 MFLX95605-14 MFLX95605-34
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Solva™, Avantor®
Hadičky Hadičky peristaltické pumpy
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attachedMore Product Information