Tisk…
Masterflex® Ismatec® Reglo Digital Gear Pump Drives with MasterflexLive®, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX78018-50-EUEA 106480 CZK
MFLX78018-50-EU
Masterflex® Ismatec® Reglo Digital Gear Pump Drives with MasterflexLive®, Avantor®
Čerpadla Pohony čerpadel Pohony zubových čerpadel
Achieve high pressure and accurate, pulseless flow -- with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • Compatible with Micropump A-mount and B-mount style gear pump heads for flow ranges from 1.7 to 4680 ml/min
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets
More Product Information