Tisk…
Masterflex® Ismatec® Reglo Digital Multichannel Pumps, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX78018-10-EUEA 94980 CZK
MFLX78018-10-EU MFLX78018-12-EU MFLX78018-14-EU MFLX78018-20-EU MFLX78018-22-EU MFLX78018-24-EU
Masterflex® Ismatec® Reglo Digital Multichannel Pumps, Avantor®
Čerpadla Čerpadla peristaltická
Versatile multichannel pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Anti-drip and speed