Tisk…
Masterflex® L/S® Variable-Speed Economy Modular Drive Systems, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX77913-02EA 54740 CZK
MFLX77913-02 MFLX77913-12 MFLX77913-07-EU MFLX77913-17-EU
Masterflex® L/S® Variable-Speed Economy Modular Drive Systems, Avantor®
Čerpadla Čerpadla peristaltická
Versatile variable-speed pump system with a wide flow range and simple controls.
  • Separate motor and controller are connected via a 6-ft (1.9 m) cable — place components where convenient or where space allows
  • Single-turn speed control and on/off switch with green power indicator — maintain speed setting while turning pump on/off
  • The 1/10-hp reversible motor has soft start and back EMF for ±2% speed control
  • Painted steel housing is IP33 rated for protection from vertically falling water
Choose a convenient, complete system all with one catalog number. AMore Product Information