Tisk…
Masterflex® L/S® Easy-Load® 3 Pump Heads for Precision Tubing, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX77800-50EA 12100 CZK
MFLX77800-50 MFLX77800-60
Masterflex® L/S® Easy-Load® 3 Pump Heads for Precision Tubing, Avantor®
Čerpadla Hlavy čerpadel Peristaltické hlavy čerpadel
Mount and stack heads without tools or hardware.
  • Flow range 0.06 to 2300 ml/min—flow rate depends on drive rpm and tubing size
  • Twist-lock mounting feature lets you mount and stack heads in seconds
  • Included mounting plate adapts Easy-Load® 3 to L/S drives — locking tab secures pump head during operation in either direction
  • Same side tubing entry/exit permits easy integration into space-limited applications — mount and operate pump head in any of four positions—depending on drive
  • Suitable for use with Masterflex® L/S® Precision tubing
The design of theMore Product Information