Tisk…
Masterflex® I/P® Easy-Load® Pump Head Replacement Main Body Assembly, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX77601-67EA 20930 CZK
MFLX77601-67 MFLX77602-07 MFLX77602-17
Masterflex® I/P® Easy-Load® Pump Head Replacement Main Body Assembly, Avantor®
Čerpadla Čerpadla peristaltická
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
  • Suitable for use with pump heads 77601-60 and 77601-62
Keep your pump head operating at its best with these replacement main body and occusion bed assemblies.