Tisk…
Masterflex® I/P® MasterSense™ Drives, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX77421-00EA 260950 CZK
MFLX77421-00 MFLX77421-02 MFLX77421-00-EU MFLX77421-02-EU
Masterflex® I/P® MasterSense™ Drives, Avantor®
Čerpadla Pohony čerpadel Pohony peristaltických čerpadel
Versatile pumps for process applications – with an intuitive touch-screen interface and advanced connectivity.
  • Powerful, space-efficient drives will accept all I/P pump head and tubing options
  • 17,8 cm (7") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Three-level user access – gives you the option of controlling who can operate the pump
More Product Information