Tisk…
Masterflex® Adhesive-Coated Microscope Slides, Avantor®
Dodavatel: Avantor Fluid Handling
75955-45 75955-47
MFLX75955-45EA 3880 CZK
MFLX75955-45 MFLX75955-47
Masterflex® Adhesive-Coated Microscope Slides, Avantor®
Sklíčka mikroskopická a krycí Sklíčka mikroskopická
Don't let your samples slip away.
  • Adhesive coatings fix cells and tissues to slides for hybridization and other staining techniques
  • Extensive quality control methods ensure each batch performs optimally
  • Coating formulations are effective without interfering with staining
Use these adhesive-coated microscope slides when sample adhesion is critical for proper staining and reliable results.
A. Gelatin is the most common adhesive coating used in histology, ideal for simple stains.
B. Poly-l-lysine is a stickier coating for more advanced immunostaining, butMore Product Information