Tisk…
Masterflex® Touch-Screen Syringe Pumps, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX74905-50EA 107730 CZK
MFLX74905-50 MFLX74905-52 MFLX74905-54 MFLX74905-56 MFLX74905-58
Masterflex® Touch-Screen Syringe Pumps, Avantor®
Čerpadla Pumpy stříkačkové
Control flow rate via the user-friendly touch-screen display.
  • Flow rates from 5 pL/min to 220 ml/min
  • Full touch-screen enables you to quickly create configurations and recall them later
  • Pump can be placed on its side to provide a smaller footprint with the touch-screen display automatically rotating
  • Accuracy of ±0.3%
These Masterflex® touch-screen syringe pumps offer user-friendly programming and control. All pumps feature an accuracy of ±0.355% and a reproducibility of ±0.05% along with output options RS-232, RS-485, USB, and 15 pin D-sub connector.
More Product Information