Tisk…
Masterflex® Close-Couple Centrifugal Pumps, Avantor®
Dodavatel: Avantor Fluid Handling
72021-11 72021-12 72021-13 72021-14 72021-23 72021-24 72021-32 72021-33 72021-34 72021-21 72021-22 72021-26 72021-31 72021-36
MFLX72021-11EA 20080 CZK
MFLX72021-11 MFLX72021-12 MFLX72021-13 MFLX72021-14 MFLX72021-23 MFLX72021-24 MFLX72021-32 MFLX72021-33 MFLX72021-34 MFLX72021-21 MFLX72021-22 MFLX72021-26 MFLX72021-31 MFLX72021-36
Masterflex® Close-Couple Centrifugal Pumps, Avantor®
Čerpadla Odstředivá čerpadla
Compact stainless steel pumps available for both DC and AC input power.
  • 316 stainless steel is ideal for corrosion resistance when pumping many liquids
  • 12 and 24 VDC versions available for many of the pumps
  • 12-foot suction lift is possible with self-priming models
  • Chemical-resistant design
The 316 SS pumps are ideal for many commercial and industrial applications where good chemical compatibility is needed for the bulk transfer of the system liquid. A carbon/ceramic/Viton® bellows mechanical seal is standard on all pumps.
The stripped wires allow forMore Product Information