Tisk…
American Machine & Tool Straight 316 SS Standard Centrifugal Pumps
Dodavatel: Avantor Fluid Handling
70761-12
MFLX70761-12EA 24400 CZK
MFLX70761-12
American Machine & Tool Straight 316 SS Standard Centrifugal Pumps
Čerpadla Odstředivá čerpadla
Investment cast stainless steel (SS) pumps are built sturdier for heavy-duty, high performance applications requiring excellent chemical resistance.
  • Ideal for low-pressure OEM and chemical industry applications
  • Rotate head in 90° increments for easy installation of tubing
  • Available with totally enclosed fan cooled (TEFC) motors
  • Chemical resistant design
These stainless steel straight centrifugal pumps are designed for continuous duty high flow and high pressure applications including chemical processing, liquid transfer, cooling, pressure boosting,More Product Information