Tisk…
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX30800-02EA 680 CZK
MFLX30800-02 MFLX30800-04 MFLX30800-06 MFLX30800-08 MFLX30800-10 MFLX45508-00 MFLX45508-01 MFLX45508-02 MFLX45508-06 MFLX45508-08 MFLX45508-12 MFLX45508-16 MFLX45508-50 MFLX45508-54
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Polypropylene, Avantor®
Fitinky a konektory
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs,
More Product Information