Tisk…
Masterflex® Female Luer to Panel Mount Barb Fitting, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX40317-00EA 450 CZK
MFLX40317-00 MFLX40317-01 MFLX40317-02 MFLX40317-03 MFLX40317-04 MFLX40317-05 MFLX40317-06 MFLX40317-07 MFLX40317-08 MFLX40317-09
Masterflex® Female Luer to Panel Mount Barb Fitting, Avantor®
Fitinky a konektory
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information