Tisk…
Masterflex® Female Luer Plug Fitting, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX40316-00EA 300 CZK
MFLX40316-00 MFLX40316-01 MFLX40316-02 MFLX40316-03 MFLX40316-04 MFLX40316-05
Masterflex® Female Luer Plug Fitting, Avantor®
Fitinky a konektory
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information