Tisk…
Masterflex® Barbed Micro2 Flow Valved Female Quick-Disconnect Fittings, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX40262-22EA 3550 CZK
MFLX40262-22 MFLX40262-23 MFLX40262-24 MFLX40262-25 MFLX40262-26 MFLX40262-27 MFLX40262-28 MFLX40262-29 MFLX40262-30 MFLX40262-31 MFLX40262-32 MFLX40262-33 MFLX40262-34 MFLX40262-35 MFLX40262-36
Masterflex® Barbed Micro2 Flow Valved Female Quick-Disconnect Fittings, Avantor®
Fitinky a konektory
Mechanical innovations reduce risk of fluid cross-over and contamination.
  • Aseptic connection and disconnection
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
  • A complete coupler requires a male and female fitting with same flow size (order separately). Compatible with Male Micro2 Quick-Disconnect fittings.
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass throughMore Product Information