Tisk…
Masterflex® 1-1/2" Sanitary Clamp High Flow Female Quick-Disconnect Fittings, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX40257-83EA 4530 CZK
MFLX40257-83 MFLX40259-00 MFLX40259-01 MFLX40259-02 MFLX40259-03 MFLX40259-04 MFLX40259-05 MFLX40259-06 MFLX40259-07 MFLX40259-08 MFLX40259-09 MFLX40259-10 MFLX40259-11 MFLX40259-12 MFLX40259-13 MFLX40259-14 MFLX40259-15 MFLX40259-16 MFLX40259-17 MFLX40259-18 MFLX40259-19 MFLX40259-20 MFLX40259-21 MFLX40259-22
Masterflex® 1-1/2" Sanitary Clamp High Flow Female Quick-Disconnect Fittings, Avantor®
Fitinky a konektory
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a High Flow Male fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design has matured, aMore Product Information