Tisk…
Masterflex® Barbed Medium Flow Male Quick-Disconnect Fittings, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX40250-62EA 710 CZK
MFLX40250-62 MFLX40250-63 MFLX40250-70 MFLX40250-71 MFLX40250-78 MFLX40250-79 MFLX40250-86 MFLX40250-87 MFLX40250-93 MFLX40250-94 MFLX40251-01 MFLX40251-02
Masterflex® Barbed Medium Flow Male Quick-Disconnect Fittings, Avantor®
Fitinky a konektory
Mechanical innovations reduce risk of fluid cross-over and contamination. A mating connection is made with a Medium Flow Female fitting.
  • Aseptic connect and disconnect
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass through at lower line pressures in a smaller body. As the overall quick disconnect design hasMore Product Information