Tisk…
Harwil In-Line Liquid Level Switches
Dodavatel: Avantor Fluid Handling
34001-55 34001-57 34001-59 34001-61 34001-63
MFLX34001-55EA 6780 CZK
MFLX34001-55 MFLX34001-57 MFLX34001-59 MFLX34001-61 MFLX34001-63
Harwil In-Line Liquid Level Switches
Senzory úrovňové
Eliminate nuisance switch tripping with a wide range of differentials.
  • Large differentials are immune to wave action and turbulence (L-21VCR models)
  • Single-point switch provides pump up/pump down level control
  • Withstands corrosive and particle-contaminated working fluids
This series of liquid level switches are easy to install, operate and maintain. Through-wall NPT process fittings and NPT electrical ports facilitate integration into existing systems. Operation checks and troubleshooting require only a multimeter.
Wetted materials are fiberglass-More Product Information